Anti-AMOTL1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16871-1-AP-150UL
10082-848EA 402 USD
10082-848
Anti-AMOTL1 Rabbit Polyclonal Antibody
Antibodies
The protein encoded by this gene is a peripheral membrane protein that is a component of tight junctions or TJs. TJs form an apical junctional structure and act to control paracellular permeability and maintain cell polarity. This protein is related to angiomotin, an angiostatin binding protein that regulates endothelial cell migration and capillary formation.

Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: AMOTL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR