Anti-AMOT Rabbit Polyclonal Antibody

Supplier: Proteintech
16870-1-AP-150UL
10082-846EA 402 USD
10082-846
Anti-AMOT Rabbit Polyclonal Antibody
Antibodies
Angiomotin belongs to the motin family of angiostatin binding protein. The encoded protein is expressed predominantly in endothelial cells of capillaries as well as larger vessels of the placenta where it may mediate the inhibitory effect of angiostatin on tube formation and the migration of endothelial cells toward growth factors during the formation of new blood vessels. Most abundant expression was found in placenta and skeletal muscle. AMOT has two isoforms with MW 130 kDa (p130) and 80 kDa (p80). This antibody recognizes p130 only.

Western Blot: Human Stomach Tissue, 1:200-1:2000

Type: Primary
Antigen: AMOT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR