Anti-FMO5 Rabbit Polyclonal Antibody

Supplier: Proteintech
16864-1-AP-150UL
10087-080EA 402 USD
10087-080
Anti-FMO5 Rabbit Polyclonal Antibody
Antibodies
Microsomal flavin-containing monooxygenases (FMOs) [dimethylaniline monooxygenase (N-oxide forming) catalyze the FAD-, NADPH- and O2-dependent oxidation of a large number of structurally diverse compounds, including drugs, pesticides, and industrial chemicals containing a soft nucleophile. FMO5, which belongs to the FMO family, is a lesser component of human liver microsomes and is present at about one-third the level of FMO3. FMO5 protein is also present at very low levels in kidney, however, FMO5 exhibits a severely restricted substrate specificity for most drugs and other xenobiotics examined to date. It has 2 isoforms produced by alternative splicing.

IHC: Human breast cancer Tissue, 1:20-1:200; Western Blot: Multi-Cells, 1:200-1:2000

Type: Primary
Antigen: FMO5-specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR