Anti-CK10 Rabbit Polyclonal Antibody

Supplier: Proteintech
16855-1-AP-150UL
10089-378EA 402 USD
10089-378
Anti-CK10 Rabbit Polyclonal Antibody
Antibodies
Keratins are a large family of proteins that form the intermediate filament cytoskeleton of epithelial cells, which are classified into two major sequence types. Type I keratins are a group of acidic intermediate filament proteins, including K9–K23, and the hair keratins Ha1–Ha8. Type II keratins are the basic or neutral courterparts to the acidic type I keratins, including K1–K8, and the hair keratins, Hb1–Hb6. As a type I keratin, keratin 10 is a suprabasal marker of differentiation in stratified squamous epithelia. This antibody is specifically against KRT10.

IHC: Human lung cancer, 1:20-1:200

Type: Primary
Antigen: CK10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR