Anti-CK1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16848-1-AP-150UL
10089-374EA 402 USD
10089-374
Anti-CK1 Rabbit Polyclonal Antibody
Antibodies
Keratins are a large family of proteins that form the intermediate filament cytoskeleton of epithelial cells, which are classified into two major sequence types. Type I keratins are a group of acidic intermediate filament proteins, including K9–K23, and the hair keratins Ha1–Ha8. Type II keratins are the basic or neutral courterparts to the acidic type I keratins, including K1–K8, and the hair keratins, Hb1–Hb6. Keratin-1 is type II keratin. It is a specific marker for terminal differentiation in mammalian epidermis. It is specifically expressed in the spinous and granular layers of the epidermis with family member KRT10 and mutations in these genes have been associated with bullous congenital ichthyosiform erythroderma. This antibody is specifically against ketatin I.

Western Blot:A375 Cells, 1:500-1:5000; IHC: Human skin Tissue, 1:20-1:200; IP:A431 Cells, 1:200-1:2000; IF:A431 Cells, 1:20-1:200

Type: Primary
Antigen: CK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR