Anti-USP13 Rabbit Polyclonal Antibody

Supplier: Proteintech
16840-1-AP-150UL
10096-840EA 402 USD
10096-840
Anti-USP13 Rabbit Polyclonal Antibody
Antibodies
USP13(Ubiquitin carboxyl-terminal hydrolase 13) is also named as ISOT3 and belongs to the peptidase C19 family. It is involved in ubiquitin-dependent protein catabolism. USP13 is responsible for MITF deubiquitination, utilizing a short hairpin RNA library against known deubiquitinating enzymes. This protein has 2 isoforms produced by alternative splicing.

Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200; IP: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: USP13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR