Anti-PIK3IP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16826-1-AP-150UL
10092-268EA 402 USD
10092-268
Anti-PIK3IP1 Rabbit Polyclonal Antibody
Antibodies
PIK3IP1(Phosphoinositide-3-kinase-interacting protein 1) is also named as HGFL. The class IA phosphoinositol-3-kinases (PI3Ks) regulate important cellular processes such as proliferation, growth, survival, motility and metabolism. PIK3IP1 is a transmembrane protein that possesses a region in its intracellular domain that shares homology with the p85 regulatory subunit of PI3K. It has 4 isoforms produced by alternative splicing. It also has a signal peptide and two glycosylation sites.

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: PIK3IP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR