Anti-GFAP Rabbit Polyclonal Antibody

Supplier: Proteintech
16825-1-AP-150UL
10081-798EA 537.7 USD
10081-798
Anti-GFAP Rabbit Polyclonal Antibody
Antibodies
GFAP (Glial fibrillary acidic protein ), an intermediate-filament (IF) protein , is specifically expressed in cells of astroglial lineage and is widely used to mark astroglia in the brain. It is also used as a marker for intracranial and intraspinal tumors arising from astrocytes.

Western Blot:C6 Cells, 1:200-1:2000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: GFAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR