Anti-SRF Rabbit Polyclonal Antibody

Supplier: Proteintech
16821-1-AP-150UL
10095-158EA 402 USD
10095-158
Anti-SRF Rabbit Polyclonal Antibody
Antibodies
Serum response factor (SRF) is a transcription factor that binds the serum response element (SRE), a sequence that mediates the transient response of many cellular genes to growth stimulation. SRF is required for cardiac differentiation and maturation due to the role in cardiac cell growth and muscle gene regulation. The calculated molecular weight of SRF is 51 kDa, but the phosphorylated SRF protein is 60-70 kDa.

Western Blot: HeLa Cells, 1:200-1:1000; IHC: Human Brain Tissue, 1:20-1:200; IP: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: SRF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR