Anti-IL1B Rabbit Polyclonal Antibody

Supplier: Proteintech
16806-1-AP-150UL
10088-702EA 537.7 USD
10088-702
Anti-IL1B Rabbit Polyclonal Antibody
Antibodies
Interleukin-1, produced mainly by blood monocytes, mediates the panoply of host reactions collectively known as acute phase response. It is identical to endogenous pyrogen. The multiple biologic activities that define IL1 are properties of a 15- to 18-kD protein that is derived from a 30- to 35-kD precursor. Interleukin 1β (IL-1B) is a member of the interleukin 1 cytokine family. It is a pro-inflammatory cytokine against infection, playing an important role in the pathogenesis of cancers. It signals through various adaptor proteins and kinases that lead to activation of numerous downstream targets. The expression level of IL-1B is very low.

Western Blot: RAW 264.7 Cells, 1:200-1:1000; IHC: Human skin Tissue, 1:20-1:200; IF: HeLa Cells, 1:20-1:200

Type: Primary
Antigen: IL1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR