Anti-LITAF Rabbit Polyclonal Antibody

Supplier: Proteintech
16797-1-AP-150UL
10089-716EA 402 USD
10089-716
Anti-LITAF Rabbit Polyclonal Antibody
Antibodies
Lipopolysaccharide-induced TNF factor(LITAF) is a transcription factor that identified as a regulator of TNF-alpha gene expression. It may has a role in protein degradation pathways and in tumor necrosis factor alpha(TNF-alpha) gene expression. Also it may regulate the expression of the CCL2/MCP-1 chemokine through NFKB1.Defects in LITAF may be involved in extramammary Paget disease (EMPD) carcinogenesis

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human spleen Tissue, 1:20-1:200

Type: Primary
Antigen: LITAF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR