Anti-MPP8 Rabbit Polyclonal Antibody

Supplier: Proteintech
16796-1-AP-150UL
10090-386EA 402 USD
10090-386
Anti-MPP8 Rabbit Polyclonal Antibody
Antibodies
The M-phase phosphoprotein 8 (MPP8) was firstly identified to coimmunoprecipitate with the RanBPM-comprised large protein complex . It also binds to methylated H3K9 and recruited the H3K9 methyltransferases GLP and ESET, as well as DNA methyltransferase 3A (DNMT3A) to the promoter of the E-cadherin gene, a key regulator of tumor cell growth and epithelial-to-mesenchymal transition (EMT) . This binding event of MPP8-H3K9me modulates the expression of E-cadherin, a key regulator of cell-cell adhesion with implications in embryonic development, tissue morphogenesis and homeostasis , together with H3K9 methyltransferases GLP and ESET and DNA methyltransferase 3a (Dnmt3a) .

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: MPP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR