Anti-PRPSAP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16790-1-AP-150UL
10092-844EA 402 USD
10092-844
Anti-PRPSAP1 Rabbit Polyclonal Antibody
Antibodies
IHC: Human cervix, 1:20-1:200

Type: Primary
Antigen: PRPSAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR