Anti-DCTD Rabbit Polyclonal Antibody

Supplier: Proteintech
16784-1-AP-150UL
10085-712EA 402 USD
10085-712
Anti-DCTD Rabbit Polyclonal Antibody
Antibodies
DCTD(Deoxycytidylate deaminase) is also named as dCMP deaminase and belongs to the cytidine and deoxycytidylate deaminase family. It catalyzes the deamination of dCMP to dUMP, thus providing the nucleotide substrate for thymidylate synthase. Control of deaminase activity at this juncture in deoxyribonucleotide metabolism is determined by the ratio of dCTP to dTTP in the cell, since the enzyme is allosterically activated by dCTP and inhibited by dTTP. It has 2 isoforms produced by alternative splicing and can exsit as a dimer.

Western Blot: Human Brain Tissue, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: DCTD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR