Anti-SPAG11A Rabbit Polyclonal Antibody

Supplier: Proteintech
16770-1-AP-150UL
10095-040EA 402 USD
10095-040
Anti-SPAG11A Rabbit Polyclonal Antibody
Antibodies
Sperm binding proteins and their C-terminal peptides of the Sperm Associated Antigen 11 (SPAG11) family are found to play an important role in epididymal innate immunity in addition to their role in sperm maturation.SPAG11 peptides resulting due to cleavage at the furin-like proprotein convertase motif are reported in the epididymal fluid, ejaculate and on sperm and their antimicrobial activity demonstrated.SPAG11A belongs to this family and is also named as EP2,HE2. It has 4 isoforms produced by alternative splicing.This antibody is specific to SPAG11.

Western Blot: Human Testis Tissue, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: SPAG11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR