Anti-IL15RA Rabbit Polyclonal Antibody

Supplier: Proteintech
16744-1-AP-150UL
10088-768EA 402 USD
10088-768
Anti-IL15RA Rabbit Polyclonal Antibody
Antibodies
Interleukin-15 (IL-15) is a pleiotropic cytokine that plays an important role in both innate and adaptive immunity. IL-15 is mainly produced by activated monocytes, macrophages and dendritic cells, and is structurally similar to IL-2 . These two cytokines share the same IL-2/15Rβ and common γ-chain receptor subunits. In addition, IL-2 and IL-15 have their own private α-chain receptor subunit, IL-2Rα and IL-15Rα, respectively . The IL-15Rα chain is expressed by many cell types including monocytes, DCs, NK, T cells and fibroblasts. Several isoforms of IL-15Rα exist and are generated either by alternative splicing or by proteolytic cleavage .

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Liver, 1:20-1:200

Type: Primary
Antigen: IL15RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR