Anti-TBX3 Rabbit Polyclonal Antibody

Supplier: Proteintech
16741-1-AP-150UL
10095-668EA 402 USD
10095-668
Anti-TBX3 Rabbit Polyclonal Antibody
Antibodies
TBX3, also named as T-box protein 3, is a 743 amino acid protein, which contains 1 T-box DNA-binding domain. TBX3 may localize in the nucleus and is widely expressed in various tissues. TBX3 as a transcriptional repressor is involved in developmental processes and probably plays a role in limb pattern formation. TBX3 plays an important role on osteogenic differentiation and proliferation of human mesenchymal stem cell derived from adipose tissue.

Western Blot: Mouse Placenta Tissue, 1:200-1:2000

Type: Primary
Antigen: TBX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR