Anti-TRMT6 Rabbit Polyclonal Antibody

Supplier: Proteintech
16727-1-AP-150UL
10096-390EA 402 USD
10096-390
Anti-TRMT6 Rabbit Polyclonal Antibody
Antibodies
Transfer RNA (tRNA) modifications help regulate the efficiency of mRNA translation by maintaining the correct reading frames within mRNA. TRMT6 (tRNA methyltransferase 6 ), also known as GCD10 or TRM6, is a 497 amino acid protein that localizes to the nucleus and exists as a heterodimer with TRM61. Expressed in liver, brain, ovary and testis, TRMT6 functions as a substrate-binding subunit of tRNA and is thought to catalyze the formation of N(1)-methyladenine at position 58 in initiator methionyl-tRNA. TRMT6 exists as three alternatively spliced isoforms that, in response to DNA damage, may be phosphorylated by ATM or ATR. This antibody is specific to 56kd TRMT6 protein.

Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human oesophagus Tissue, 1:20-1:200

Type: Primary
Antigen: TRMT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR