Anti-Coproporphyrinogen oxidase Rabbit Polyclonal Antibody

Supplier: Proteintech
16724-1-AP-150UL
10082-048EA 402 USD
10082-048
Anti-Coproporphyrinogen oxidase Rabbit Polyclonal Antibody
Antibodies
CPOX(Coproporphyrinogen-III oxidase, mitochondrial) is also named as CPO and CPX. Human CPO is a 76 kDa protein that is active as a homodimer in the mitochondrial intermembrane pace.Mutations in human CPO gene predict the clinical outcome of the disease, with either hepatic hereditary coproporphyria or hematological manifestations of erythropoietic harderoporphyria.CPOX has a transit peptide of 110 aa and the chain contains 344 aa.

Western Blot:K-562 Cells, 1:1000-1:10000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: Coproporphyrinogen oxidase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR