Anti-NPL Rabbit Polyclonal Antibody

Supplier: Proteintech
16715-1-AP-150UL
10091-256EA 402 USD
10091-256
Anti-NPL Rabbit Polyclonal Antibody
Antibodies
The N-acetylneuraminate lyase(NPL) from human (hNAL) which consists of 320 amino acids, has a molecular mass of 35.2 kDa and it catalyses the reversible aldol reaction of sialic acid, and has been extensively used in the synthesis of sialic acids and its analogues. The enzyme is widely distributed and are present in numerous pro- and eukaryotic cells, in which they are localized only in the cytosol. It belongs to the DHDPS family and has 5 isoforms produced by alternative splicing.

Western Blot: Mouse Kidney Tissue, 1:200-1:2000; IF: MCF-7 Cells, 1:10-1:100

Type: Primary
Antigen: NPL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR