Anti-CYP20A1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16702-1-AP-150UL
10085-506EA 402 USD
10085-506
Anti-CYP20A1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:1000-1:10000; IHC: Human Liver cancer, 1:20-1:200

Type: Primary
Antigen: CYP20A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR