Anti-IDI2 Rabbit Polyclonal Antibody

Supplier: Proteintech
16701-1-AP-150UL
10088-580EA 402 USD
10088-580
Anti-IDI2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Skeletal Muscle Tissue, 1:1000-1:10000

Type: Primary
Antigen: IDI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR