Anti-NKD2 Rabbit Polyclonal Antibody

Supplier: Proteintech
16699-1-AP-150UL
10091-124EA 402 USD
10091-124
Anti-NKD2 Rabbit Polyclonal Antibody
Antibodies
Naked2 (protein naked cuticle homolog 2), encoded by NKD2 gene, was identified as a cell autonomous antagonist of the canonical Wnt signaling pathway. Naked2 acts as a cargo recognition and targeting protein to ensure proper delivery and fusion of TGF-alpha-containing vesicles to a distinct region at the basolateral surface of polarized epithelial cells . EGFR-independent action of TGF-alpha protects Naked2 from proteasomal degradation, thus ensuring its delivery to the basolateral surface . Myristoylated Naked2 antagonizes Wnt-beta-catenin activity possibly by degrading Dishevelled-1 at the plasma membrane .

Western Blot: Mouse Kidney Tissue, 1:200-1:1000

Type: Primary
Antigen: NKD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR