Anti-CA13 Rabbit Polyclonal Antibody

Supplier: Proteintech
16696-1-AP-150UL
10083-912EA 402 USD
10083-912
Anti-CA13 Rabbit Polyclonal Antibody
Antibodies
Carbonic anhydrases (CAs) are zinc-containing metalloenzymes that catalyze reversible hydration of carbon dioxide. The mammalian α-CA gene family has been reported to include at least eleven enzymatically active isoforms with different structural and catalytic properties. Four of the active CA isozymes are cytosolic (CA I, II, III, and VII), four are membrane-associated(CA IV, IX, XII, and XIV), two are mitochondrial (CA VA and VB), and one is a secretory form (CA VI). CA13 is a cytosolic proteinmay that may play a role in embryogenesis and perturbation of its function by genetic modification could potentially lead to developmental abnormalities.

Western Blot:NIH/3T3 Cells, 1:500-1:5000; IHC: Human Placenta Tissue, 1:20-1:200

Type: Primary
Antigen: CA13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR