Anti-AMFR Rabbit Polyclonal Antibody

Supplier: Proteintech
16675-1-AP-150UL
10082-832EA 524.48 USD
10082-832
Anti-AMFR Rabbit Polyclonal Antibody
Antibodies
AMFR(Autocrine motility factor receptor) is also named as RNF45,gp78.It is the receptor of Autocrine motility factor(AMF),which is a protein secreted by tumor cells that stimulates tumor motility. AMFR has a potential N-glycosylation site, and several potential O-glycosylation sites.In normal fibroblasts,the expression of AMFR appeared to be regulated by cell-cell contact.

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human ovary Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP: HepG2 Cells, 1:200-1:2000

Type: Primary
Antigen: AMFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR