To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Heparanase
- Antigen Symbol:HPSE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Antigen Synonyms:HSE1.|HPSE|HPSE 1|heparanase|HPR 1|Hpa1|Endo-glucoronidase|HEP|Hpa 1|heparanase 1|HPR1|Heparanase 50 kDa subunit|HPSE1|heparanase-1|Heparanase1|HPSE_HUMAN|HPA|Heparanase 1|heparanase1|Endo glucoronidase|HSE 1
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. No.:10081-850
- Supplier no.:16673-1-AP
HPSE(Heparanase) is also named as HEP, HPA, HPA1, HPR1, HPSE1, HSE1 and belongs to the glycosyl hydrolase 79 family. It is a endoglycosidase that cleaves heparan sulfate proteoglycans (HSPGs) into heparan sulfate side chains and core proteoglycans. HPSE is essential in the disassembly of the extracellular matrix (ECM) by invading cells. It has 3 isoforms produced by alternative splicing with the molecular weight of 61 kDa, 55 kDa and 53 kDa. The full length protein has six glycosylation sites. The cleavage of the 65 kDa form leads to the generation of a linker peptide, and 8 kDa and 50 kDa products. The active form, the 8/50 kDa heterodimer, is resistant to degradation and glycosylation of the 50 kDa subunit appears to be essential for its solubility.
Western Blot:DU 145 Cells, 1:200-1:1000; IHC: Human Liver cancer Tissue, 1:20-1:200
Type: Primary
Antigen: HPSE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat