Anti-CD3D Rabbit Polyclonal Antibody

Supplier: Proteintech
16669-1-AP-150UL
10084-312EA 402 USD
10084-312
Anti-CD3D Rabbit Polyclonal Antibody
Antibodies
CD3 is a complex of proteins that directly associates with the T cell receptor (TCR). The TCR/CD3 complex of T-lymphocytes consists of either a TCR alpha/beta or TCR gamma/delta heterodimer coexpressed at the cell surface with the invariant subunits of CD3 labeled gamma, delta, epsilon, zeta, and eta. The TCR recognizes antigens bound to major histocompatibility complex (MHC) molecules. TCR-mediated peptide-MHC recognition is transmitted to the CD3 complex, leading to the intracellular signal transduction. CD3 is considered to be a pan-T cell marker for detection of normal and neoplastic T cells.

Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human Lymphoma Tissue, 1:20-1:200; FC:Jurkat Cells, N/A

Type: Primary
Antigen: CD3D
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR