Anti-NDUFA5 Rabbit Polyclonal Antibody

Supplier: Proteintech
16640-1-AP-150UL
10090-634EA 402 USD
10090-634
Anti-NDUFA5 Rabbit Polyclonal Antibody
Antibodies
NDUFA5, also named as CI-13kD-B and NDUA5, belongs to the complex I NDUFA5 subunit family. It is accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) which functions in the transfer of electrons from NADH to the respiratory chain. NDUFA5,an enrichment of mitochondrial protein, can be used as a mitochondrial marker protein.

Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: NDUFA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR