Anti-COPS7A Rabbit Polyclonal Antibody

Supplier: Proteintech
16628-1-AP-150UL
10085-014EA 402 USD
10085-014
Anti-COPS7A Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Heart Tissue, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200; IP: Mouse Heart Tissue, 1:200-1:1000

Type: Primary
Antigen: COPS7A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR