Anti-ME1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16619-1-AP-150UL
10090-074EA 402 USD
10090-074
Anti-ME1 Rabbit Polyclonal Antibody
Antibodies
ME1(NADP-dependent malic enzyme) belongs to the malic enzymes family. This malic enzyme catalyzes the reversible oxidative decarboxylation of malate and is a link between the glycolytic pathway and the citric acid cycle. The reaction is L-malate plus NADP(+) to form pyruvate, CO(2), and NADPH. The predicted protein contains 572 amino acids and has a calculated molecular mass of 64.1 kDa.

Western Blot: Mouse Liver Tissue, 1:500-1:5000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: ME1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR