Anti-ZNF143 Rabbit Polyclonal Antibody

Supplier: Proteintech
16618-1-AP-150UL
10097-304EA 402 USD
10097-304
Anti-ZNF143 Rabbit Polyclonal Antibody
Antibodies
Zinc finger protein143(ZNF143) contains a separete acidic activation and 7 zinc finger domains. It's a transcription factor that required for activation of the majority of vertebrate snRNA and snRNA-type genes, which transcribed by RNA pol II and pol III. Also, it binds to the SPH motif of small nuclear RNA (snRNA) gene promoters and participates in efficient U6 RNA polymerase III transcription via its interaction with CHD8

Western Blot: HepG2 Cells, 1:500-1:5000

Type: Primary
Antigen: ZNF143
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR