Anti-NUMA Rabbit Polyclonal Antibody

Supplier: Proteintech
16607-1-AP-150UL
10091-394EA 524.48 USD
10091-394
Anti-NUMA Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:200-1:1000

Type: Primary
Antigen: NUMA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR