Anti-OTUD7B Rabbit Polyclonal Antibody

Supplier: Proteintech
16605-1-AP-150UL
10091-638EA 402 USD
10091-638
Anti-OTUD7B Rabbit Polyclonal Antibody
Antibodies
OUTD7B is a member of the A20 family of the deubiquitinating cysteine proteases, which has deubiquitinating activity that is directed towards 'Lys-48' or 'Lys-63'-linked polyubiquitin chains. It plays a key role in the negative feedback regulation of NF-κB signaling in response to multiple stimuli and has been described as central gatekeeper in inflammation and immunity.

Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200; IP:Jurkat Cells, 1:200-1:2000

Type: Primary
Antigen: OTUD7B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR