Anti-LIPH Rabbit Polyclonal Antibody

Supplier: Proteintech
16602-1-AP-150UL
10089-712EA 402 USD
10089-712
Anti-LIPH Rabbit Polyclonal Antibody
Antibodies
LIPH(Lipase member H) is also named as LPDLR, MPAPLA1, PLA1B. It is a phosphatidic acid-selective phospholipase A1 (PLA1) that produces 2-acyl lysophosphatidic acid (LPA) and belongs to the AB hydrolase superfamily and Lipase family.LIPH has a 12-residue lid region, which likely covers a catalytic pocket, and 4 potential N-linked glycosylation sites. Defects in LIPH are the cause of hypotrichosis type 7 (HYPT7).

Western Blot:A549 Cells, 1:200-1:2000; IHC: Human pancreas Tissue, 1:20-1:200

Type: Primary
Antigen: LIPH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR