Anti-PGCP Rabbit Polyclonal Antibody

Supplier: Proteintech
16601-1-AP-150UL
10092-144EA 402 USD
10092-144
Anti-PGCP Rabbit Polyclonal Antibody
Antibodies
PGCP(plasma glutamate carboxypeptidase) is also named as LCH1,CPQ and belongs to the peptidase M28 family. It may play an important role in the hydrolysis of circulating peptides. It catalyzes more efficiently the hydrolysis of dipeptides with unsubstituted terminals into amino acids. It is a proteinase that acts on the unsubstituted N- and C-termini of dipeptides. It has been suggested that this PGCP is involved in the release of thyroxine. Human PGCP is a metallopeptidase with five potential glycosylation sites, whose activity depends on dimerizationand it can be detected a band of 65-72kd in FRTL-5 cells which is pro-PGCP. It can be N-glycosylated. It has the pro-form of 60-65 kDa and mature form of 55 kDa.

Western Blot:L02 Cells, 1:200-1:2000

Type: Primary
Antigen: PGCP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR