Anti-MED31 Rabbit Polyclonal Antibody

Supplier: Proteintech
16590-1-AP-150UL
10090-100EA 402 USD
10090-100
Anti-MED31 Rabbit Polyclonal Antibody
Antibodies
MED31, also named as SOH1 or CGI-125, is a 131 amino acid protein, which belongs to the Mediator complex subunit 31 family. MED31 localizes in the nucleus. MED31 as a component of the Mediator complex, a coactivator is involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. MED31 functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery.

Western Blot: Mouse Testis Tissue, 1:500-1:5000

Type: Primary
Antigen: MED31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR