Anti-Pyruvate Carboxylase Rabbit Polyclonal Antibody

Supplier: Proteintech
16588-1-AP-150UL
10091-898EA 402 USD
10091-898
Anti-Pyruvate Carboxylase Rabbit Polyclonal Antibody
Antibodies
PC(pyruvate carboxylase) is a member of the family of biotin-dependent carboxylases and is found widely among eukaryotic tissues and in many prokaryotic species. It catalyses the ATP-dependent carboxylation of pyruvate to form oxaloacetate which may be utilised in the synthesis of glucose, fat, some amino acids or their derivatives and several neurotransmitters. Diabetes and hyperthyroidism increase the level of expression of pyruvate carboxylase in the long term, while its activity in the short term is controlled by the intramitochondrial concentrations of acetyl-CoA and pyruvate.

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human Liver Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: Pyruvate Carboxylase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR