Anti-SDR39U1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16585-1-AP-150UL
10094-240EA 402 USD
10094-240
Anti-SDR39U1 Rabbit Polyclonal Antibody
Antibodies
SDR39U1(Epimerase family protein SDR39U1) is also named as C14orf124, HCDI and belongs to the sugar epimerase family and SDR39U1 subfamily. It shares more sequence similarity with the sugar epimerase family than with the short-chain dehydrogenases/reductases (SDR) family. It has 3 isoforms produced by alternative splicing with the molecular weight of 35 kDa, 31 kDa and 21 kDa.

Western Blot: Human adrenal gland Tissue, 1:200-1:2000

Type: Primary
Antigen: SDR39U1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR