Anti-MRPL3 Rabbit Polyclonal Antibody

Supplier: Proteintech
16584-1-AP-150UL
10089-682EA 402 USD
10089-682
Anti-MRPL3 Rabbit Polyclonal Antibody
Antibodies
MRPL3, also named as 39S ribosomal protein L3, mitochondrial or RPML3, is a 348 amino acid protein, which belongs to the ribosomal protein L3P family. MRPL3 localizes in the Mitochondrion and has an association with combined oxidative phosphorylation deficiency 9. MRPL3 plays role in the mitochondrial proteins biosynthesis and ribosomal structure and biogenesis.

Western Blot: HeLa Cells, 1:1000-1:10000; IHC: Human cervical cancer Tissue, 1:20-1:200

Type: Primary
Antigen: MRPL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR