Anti-CK4 Rabbit Polyclonal Antibody

Supplier: Proteintech
16572-1-AP-150UL
10085-630EA 402 USD
10085-630
Anti-CK4 Rabbit Polyclonal Antibody
Antibodies
Keratins are a large family of proteins that form the intermediate filament cytoskeleton of epithelial cells, which are classified into two major sequence types. Type I keratins are a group of acidic intermediate filament proteins, including K9–K23, and the hair keratins Ha1–Ha8. Type II keratins are the basic or neutral courterparts to the acidic type I keratins, including K1–K8, and the hair keratins, Hb1–Hb6. Keratin 4 is a type II cytokeratins. It is specifically found in differentiated layers of the mucosal and esophageal epithelia together with keratin 13.

Western Blot:A431 Cells, 1:200-1:2000; IHC: Human cervix Tissue, 1:20-1:200

Type: Primary
Antigen: CK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR