Anti-Fetuin-A Rabbit Polyclonal Antibody

Supplier: Proteintech
16571-1-AP-150UL
10086-952EA 402 USD
10086-952
Anti-Fetuin-A Rabbit Polyclonal Antibody
Antibodies
Fetuin-A (alpha-2-HS-glycoprotein, AHSG), a liver borne plasma protein, contributes to the prevention of soft tissue calcification, modulates inflammation, reduces insulin sensitivity and fosters weight gain following high fat diet or ageing. It is involved in several functions, such as endocytosis, brain development and the formation of bone tissue. The protein is commonly present in the cortical plate of the immature cerebral cortex and bone marrow hemopoietic matrix, and it has therefore been postulated that it participates in the development of the tissues.

IHC: Human Liver Tissue, 1:20-1:200; Western Blot: Human Liver Tissue, 1:200-1:2000

Type: Primary
Antigen: Fetuin-A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR