Anti-TMEM11 Rabbit Polyclonal Antibody

Supplier: Proteintech
16564-1-AP-150UL
10096-078EA 402 USD
10096-078
Anti-TMEM11 Rabbit Polyclonal Antibody
Antibodies
TMEM11, also named as C17orf35 and PM1, belongs to the TMEM11 family. TMEM11 is a mitochondrial inner-membrane protein facing the matrix. It regulates mitochondrial morphogenesis independently of the DRP1/MFN fission/fusion pathways.

Western Blot: Human Placenta Tissue, 1:500-1:5000

Type: Primary
Antigen: TMEM11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR