Anti-GLI4 Rabbit Polyclonal Antibody

Supplier: Proteintech
16551-1-AP-150UL
10087-580EA 402 USD
10087-580
Anti-GLI4 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:200-1:1000

Type: Primary
Antigen: GLI4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR