Anti-CYP2C8 Rabbit Polyclonal Antibody

Supplier: Proteintech
16546-1-AP-150UL
10085-516EA 402 USD
10085-516
Anti-CYP2C8 Rabbit Polyclonal Antibody
Antibodies
CYP2C8(Cytochrome P450 2C8) is also named as CYPIIC8, cytochrome P450 IIC2, cytochrome P450 MP-12, cytochrome P450 MP-20, cytochrome P450 form 1 and S-mephenytoin 4-hydroxylase. It belongs to the cytochrome P450 family. It is a monooxygenase involved in an NADPH-dependent electron transport pathway and a hepatic drug-metabolizing enzymes that oxidizes therapeutic drugs such as taxol and cerivastatin and endobiotics such as retinoic acid and arachidonic acid. It has 2 isoforms produced by alternative splicing.

IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: CYP2C8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR