Anti-CTHRC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16534-1-AP-150UL
10085-310EA 537.7 USD
10085-310
Anti-CTHRC1 Rabbit Polyclonal Antibody
Antibodies
Collagen triple helix repeat-containing protein 1 (CTHRC1), also known protein NMTC1, is a glycosylated secreted protein. CTHRC1 was identified as a novel molecular in injured and diseased arteries where it may contribute to vascular remodeling by limiting collagen matrix deposition and promoting cell migration . It has been reported that CTHRC1 is a positive regulator of osteoblastic bone formation . Besides, studies reveal that CTHRC1 expression is implicated in various malignancies, including hepatocellular carcinoma, gastric carcinoma, colon cancer, and lung cancer .

Western Blot:A375 Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: CTHRC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR