Anti-PALMD Rabbit Polyclonal Antibody

Supplier: Proteintech
16531-1-AP-150UL
10091-774EA 402 USD
10091-774
Anti-PALMD Rabbit Polyclonal Antibody
Antibodies
Palmdelphin, encoded by PALMD gene, is a newly identified cytosolic isoform of paralemmin-1, a lipid raft-associated protein implicated in cell shape control. Paralemmin (Palm) is a prenyl-palmitoyl anchored membrane protein that can drive membrane and process formation in neurons. Previous finding suggest that palmdelphin may peripherally associate with endomembranes or cytoskeleton-linked structures. Palmdelphin appears to be 80 kDa in Western blotting, larger than the predicated of 63 kDa, and this may be due to the acidic nature of this protein or its posttranslational modification.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Liver Tissue, 1:20-1:200; IP: Mouse Heart Tissue, 1:200-1:2000

Type: Primary
Antigen: PALMD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR