Anti-NGDN Rabbit Polyclonal Antibody

Supplier: Proteintech
16524-1-AP-150UL
10091-084EA 402 USD
10091-084
Anti-NGDN Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human cerebellum Tissue, 1:200-1:2000; IHC: Human epididymis, 1:20-1:200; IF: Hela, 1:20-1:200

Type: Primary
Antigen: NGDN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR