Anti-PMEPA1 Rabbit Polyclonal Antibody

Supplier: Proteintech
16521-1-AP-150UL
10092-286EA 402 USD
10092-286
Anti-PMEPA1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PMEPA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR