Anti-DES Rabbit Polyclonal Antibody

Supplier: Proteintech
16520-1-AP-150UL
10085-774EA 537.7 USD
10085-774
Anti-DES Rabbit Polyclonal Antibody
Antibodies
Desmin is the main intermediate filament protein in skeletal and cardiac muscle cells and is essential for both the structural integrity and the survival of muscle cells. As an abundant muscle-specific protein, desmin has been widely used as a marker of muscle derived tumors. Anti-desmin is also valuable in the differential diagnosis of tumors of uncertain origin.

Western Blot: Mouse Heart Tissue, 1:1000-1:10000; IF: HepG2 Cells, 1:20-1:200; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: DES
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR