Anti-RPL31 Rabbit Polyclonal Antibody

Supplier: Proteintech
16497-1-AP-150UL
10093-718EA 402 USD
10093-718
Anti-RPL31 Rabbit Polyclonal Antibody
Antibodies
The mammalian ribosome comprises 79 ribosomal proteins and four rRNAs, which combine in equimolar ratios to form the small (40S) and large (60S) subunits. Ribosome proteins are a direct and critical target of the PI3K pathway in promoting growth.. Ribosomal protein L31 gene is a component of the 60S large ribosomal subunit encoded by RPL31 gene, while ribosomal protein L31 (RPL31) is an important constituent of peptidyltransferase center .

Western Blot: HepG2 Cells, 1:200-1:2000

Type: Primary
Antigen: RPL31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR